2,3,5-Trichlorophenol
2,3,5-Trichlorophenol
EHERC17773500
- SMILES: Oc1cc(Cl)cc(Cl)c1Cl Inchi: InChI=1S/C6H3Cl3O/c7-3-1-4(8)6(9)5(10)2-3/h1-2,10H IUPAC: 2,3,5-trichlorophenol
- Packing group: III
- Description:2,3,5-Trichlorophenol
- UN No.:2020
- Cat. No.:EHERC17773500
- Formula:C₆H₃Cl₃O
- Storage conditions:20 ℃
- Hazard class description:GHS05, GHS07, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Phenol, 2,3,5-trichloro-; 2,3,5-Trichlorophenol
- Molecular weight:197,45
- CAS No.:933-78-8
- Shipping conditions:RT



