Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

Styrene

Styrene
Catalog # EHERC16982000
Supplier:   EHRENSTORFER
Styrene
Catalog # EHERC16982000
Supplier:  EHRENSTORFER
CAS Number:  
Styrene
Styrene

Styrene

CAS Number:  
Price
39,50 €
Log in for availability
1 * 1 g
Product Details & Documents
  • SMILES: C=Cc1ccccc1
  • Inchi: InChI=1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2
  • IUPAC: styrene
  • Packing group: III

 

Specifications

Specifications

  • Description:
    Styrene
  • UN No.:
    2055
  • Cat. No.:
    EHERC16982000
  • Formula:
    C₈H₈
  • Storage conditions:
    4 °C
  • Hazard class description:
    GHS02, GHS07, GHS08, GHS09
  • GHS signal:
    Danger
  • Synonym(s):
    Styrene;Ethenylbenzene;Cinnamene;Maomin SM;NSC 62785;Phenethylene;Phenylethene;Phenylethylene;Styrol;Styrole;Styrolene;Styropol SO;TTB 7302;Vinylbenzene;Vinylbenzol;
  • Molecular weight:
    104,15
  • CAS No.:
    100-42-5
  • Shipping conditions:
    Ice pack (–20 °C)

Specifications