Methacrifos
Methacrifos
EHERC14970000
- SMILES: COC(=O)\C(=C\OP(=S)(OC)OC)\C
- Inchi: InChI=1S/C7H13O5PS/c1-6(7(8)9-2)5-12-13(14,10-3)11-4/h5H,1-4H3/b6-5+
- IUPAC: methyl (E)-3-dimethoxyphosphinothioyloxy-2-methylprop-2-enoate
- Packing group: III
- Description:Methacrifos
- UN No.:3082
- Cat. No.:EHERC14970000
- Formula:C₇H₁₃O₅PS
- Storage conditions:–18 °C
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Synonym(s):2-Propenoic acid;3-[(dimethoxyphosphinothioyl)oxy]-2-methyl-;methyl ester;(2E)-;2-Propenoic acid;3-[(dimethoxyphosphinothioyl)oxy]-2-methyl-;methyl ester;(E)-;CGA 20168;trans-Methacrifos;Methacrifos
- Molecular weight:240,21
- CAS No.:62610-77-9
- Shipping conditions:RT



