Fluazifop-methyl
Fluazifop-methyl
EHERC13670400
- SMILES: COC(=O)C(C)Oc1ccc(Oc2ccc(cn2)C(F)(F)F)cc1
- Inchi: InChI=1S/C16H14F3NO4/c1-10(15(21)22-2)23-12-4-6-13(7-5-12)24-14-8-3-11(9-20-14)16(17,18)19/h3-10H,1-2H3
- IUPAC: methyl 2-[4-[5-(trifluoromethyl)pyridin-2-yl]oxyphenoxy]propanoate
- Packing group: III
- Description:Fluazifop-methyl
- UN No.:3077
- Cat. No.:EHERC13670400
- Formula:C₁₆H₁₄F₃NO₄
- Storage conditions:20 °C
- Hazard class description:GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Fluazifop methyl ester;SL 236;Fluazifop-methyl;Fluazifop Methyl
- Molecular weight:341,28
- CAS No.:69335-90-6
- Shipping conditions:RT



