Physcion
Physcion
Catalog # EHERC16192000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: COc1cc(O)c2C(=O)c3c(O)cc(C)cc3C(=O)c2c1
- Inchi: InChI=1S/C16H12O5/c1-7-3-9-13(11(17)4-7)16(20)14-10(15(9)19)5-8(21-2)6-12(14)18/h3-6,17-18H,1-2H3
- IUPAC: 1,8-dihydroxy-3-methoxy-6-methylanthracene-9,10-dione
Specifications
Specifications
- Description:Physcion
- Cat. No.:EHERC16192000
- Formula:C₁₆H₁₂O₅
- Storage conditions:4 °C
- Hazard class description:GHS07
- GHS signal:Warning
- Synonym(s):Physcion; Physcion
- Molecular weight:284,26
- CAS No.:521-61-9
- Shipping conditions:RT



