Mapenterol hydrochloride
Mapenterol hydrochloride
EHERC14754000
- SMILES: Cl.CCC(C)(C)NCC(O)c1cc(Cl)c(N)c(c1)C(F)(F)F
- Inchi: InChI=1S/C14H20ClF3N2O.ClH/c1-4-13(2,3)20-7-11(21)8-5-9(14(16,17)18)12(19)10(15)6-8;/h5-6,11,20-21H,4,7,19H2,1-3H3;1H
- IUPAC: 1-[4-amino-3-chloro-5-(trifluoromethyl)phenyl]-2-(2-methylbutan-2-ylamino)ethanol;hydrochloride
- Description:Mapenterol hydrochloride
- Cat. No.:EHERC14754000
- Formula:C₁₄H₂₀ClF₃N₂O·ClH
- Storage conditions:4 °C
- Hazard class description:GHS07
- GHS signal:Warning
- Synonym(s):Mapenterol Hydrochloride; 4-Amino-3-chloro-α-[[(1,1-dimethylpropyl)amino]methyl]-5-(trifluoromethyl)benzenemethanol Hydrochloride;
- Molecular weight:361,23
- CAS No.:54238-51-6
- Shipping conditions:RT



