Orders LinkContactUsLinkComponent
Spermidine ≥98%
Spermidine ≥98%
Bottle
Catalog # APOSBUP17513-100MG
New Product
Supplier:   Apollo Scientific
Spermidine ≥98%
Catalog # APOSBUP17513-100MG
Supplier:  Apollo Scientific
New Product
CAS Number:  124-20-9
Spermidine ≥98%
Bottle
CAS Number:  124-20-9
Price
57,90 €
Log in for availability
1 * 100 mg
Product Details & Documents
Usage: Biochemical/life science applications
Synonyms:  N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine
  • CAS Number:124-20-9
  • Formula:C₇H₁₉N₃
  • Molecular Weight:145,25 g/mol
Specifications
Specifications
  • Pack type:
    Bottle
Specifications