Orders LinkContactUsLinkComponent
Estrone ≥98%
Estrone ≥98%
New Product
Supplier:   Apollo Scientific
Estrone ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
3 Options Available Below
Product Details & Documents
Usage: Biochemical/life science applications.
  • CAS Number:53-16-7
  • Formula:C₁₈H₂₂O₂
  • Molecular Weight:270,37 g/mol
Product Family Options
Product Information
  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 59,40 €
    59,40 €
    1 * 500 mg
       59,40 €
    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 71,00 €
    71,00 €
    1 * 1 g
       71,00 €
    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 194,00 €
    194,00 €
    1 * 10 g
       194,00 €
    Specifications:
    New Product