Orders LinkContactUsLinkComponent
Hydroxyectoin 95%
New Product
Supplier:   Apollo Scientific
Hydroxyectoin 95%
Supplier:  Apollo Scientific
New Product
Certificates
Hydroxyectoin 95%
4 Options Available Below
Product Details & Documents
Usage: Organic building block/reagent.
  • CAS Number:165542-15-4
  • Formula:C₆H₁₀N₂O₃
  • Molecular Weight:158,16 g/mol
Product Family Options
Product Information
  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 26,10 €
    26,10 €
    1 * 100 mg
       26,10 €
    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 37,70 €
    37,70 €
    1 * 250 mg
       37,70 €
    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 69,50 €
    69,50 €
    1 * 1 g
       69,50 €
    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 341,00 €
    341,00 €
    1 * 5 g
       341,00 €
    Specifications:
    New Product