Transport Trolley, Automatic
Transport trolley with height-adjustable design, 662×1156×923 mm, 270 lbs., Stainless steel
Catalog # MIEL11621520
New Product
Supplier: MIELE
Jump to:
Product Details & Documents
Transport trolley, automatic for loading and unloading large-scale dishwashers with electric height adjustment.
Enables ergonomic working
Infinite electric height adjustment
Also, 3 fixed, freely selectable height positions can be saved
Flexible operation thanks to highperformance battery
Installed with a removable drip tray
Battery included, the charger must be ordered separately
Specifications
- Colour:Stainless steel
- Description:Transport trolley with height-adjustable design
- W×D×H:662×1156×923 mm
- Weight:270 lbs.