Orders LinkContactUsLinkComponent
eXtreme/FV® Filter Vials
Supplier:   THOMSON INSTRUMENT
eXtreme/FV® Filter Vials
Supplier:  THOMSON INSTRUMENT
Certificates
eXtreme/FV® Filter Vials
eXtreme/FV® Filter Vials
12 Options Available Below
Product Details & Documents
Thomson's filtration products are engineered and optimised to increase the efficiency of sample preparation without compromising the results.
Thomson eXtreme/FV® Filter vials offer tiered filtration, and are designed to speed up sample prep and analysis on particulate laden samples. Save on costly repairs and service issues while eliminating multiple syringe filters, syringes, HPLC vials, and HPLC caps. The eXtreme/FV® Filter Vials are easy to press down with hands or toggle press.
These filter vials are manufactured with attention to detail and high-grade quality to ensure performance at optimal levels. Filter vials find a wide variety of applications ranging from life sciences, drug discovery, monitoring, and analysis. They are universal for all different types of autosamplers, including UHPLC.
Product Family Options
Product Information
  • Membrane
    Pore size
    Colour
    Availability
    Price
  • THMS85530-200
    eXtreme/FV® Filter vial
    eXtreme/FV® Filter vial
    PTFE, Tiered filtration
    0,2 µm
    Green
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items - 875,00 €
    875,00 €
    1 * 200 items
       875,00 €
  • THMS85530-500
    eXtreme/FV® Filter vial
    eXtreme/FV® Filter vial
    PTFE, Tiered filtration
    0,2 µm
    Green
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - 2 190,00 €
    2 190,00 €
    1 * 500 items
       2 190,00 €
  • THMS85540-200
    eXtreme/FV® Filter vial
    eXtreme/FV® Filter vial
    PTFE, Tiered filtration
    0,2 µm
    Blue
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items - 875,00 €
    875,00 €
    1 * 200 items
       875,00 €
  • THMS85540-500
    eXtreme/FV® Filter vial
    eXtreme/FV® Filter vial
    PTFE, Tiered filtration
    0,45 µm
    Blue
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - 2 190,00 €
    2 190,00 €
    1 * 500 items
       2 190,00 €
  • THMS85541-200
    eXtreme/FV® Filter vial
    eXtreme/FV® Filter vial
    PVDF, Tiered filtration
    0,2 µm
    Yellow
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items - 875,00 €
    875,00 €
    1 * 200 items
       875,00 €
  • THMS85541-500
    eXtreme/FV® Filter vial
    eXtreme/FV® Filter vial
    PVDF, Tiered filtration
    0,45 µm
    Yellow
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - 2 190,00 €
    2 190,00 €
    1 * 500 items
       2 190,00 €
  • THMS85538-200
    eXtreme/FV® Filter vial
    eXtreme/FV® Filter vial
    Nylon, Tiered filtration
    0,2 µm
    Black
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items - 875,00 €
    875,00 €
    1 * 200 items
       875,00 €
  • THMS85538-500
    eXtreme/FV® Filter vial
    eXtreme/FV® Filter vial
    Nylon, Tiered filtration
    0,2 µm
    Black
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - 2 190,00 €
    2 190,00 €
    1 * 500 items
       2 190,00 €
  • THMS85539-200
    eXtreme/FV® Filter vial
    eXtreme/FV® Filter vial
    Nylon, Tiered filtration
    0,45 µm
    Pink
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items - 875,00 €
    875,00 €
    1 * 200 items
       875,00 €
  • THMS85539-500
    eXtreme/FV® Filter vial
    eXtreme/FV® Filter vial
    Nylon, Tiered filtration
    0,45 µm
    Pink
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - 2 190,00 €
    2 190,00 €
    1 * 500 items
       2 190,00 €
  • THMS85535-200
    eXtreme/FV® Filter vial
    eXtreme/FV® Filter vial
    PES, Tiered filtration
    0,2 µm
    Grey
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items - 875,00 €
    875,00 €
    1 * 200 items
       875,00 €
  • THMS85535-500
    eXtreme/FV® Filter vial
    eXtreme/FV® Filter vial
    PES, Tiered filtration
    0,2 µm
    Grey
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - 2 190,00 €
    2 190,00 €
    1 * 500 items
       2 190,00 €