Order Entry
Slovenia
Orders LinkContactUsLinkComponent
FH535 ≥98% Wnt/beta-cantenin inhibitor

FH535 ≥98% Wnt/beta-cantenin inhibitor

Supplier:   Abcam
FH535 ≥98% Wnt/beta-cantenin inhibitor
Supplier:  Abcam
Certificates

2 Options Available Below

Product Details & Documents

Wnt/β-cantenin inhibitor and PPARγ and PPARδ antagonist. Suppresses proliferation of cancer cells in vitro. Antitumor agent.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  2,5-Dichloro-N-(2-methyl-4-nitrophenyl)benzenesulphonamide
  • CAS Number:108409-83-2
  • Molecular Weight:361,2 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% 2,5-Dichloro-N-(2-methyl-4-nitrophenyl)-benzenesulfonamide
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 134,00 €
    134,00 €
    1 * 10 mg
       134,00 €

  • Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 487,00 €
    487,00 €
    1 * 50 mg
       487,00 €