ContactUsLinkComponent
Soft Care Des E H5 Alcohol based Hand Disinfectant
Soft Care Des E H5 Alcohol based Hand Disinfectant
Catalog # DIVY101106601
Supplier:  Diversey
New Product
Soft Care Des E H5 Alcohol based Hand Disinfectant
Catalog # DIVY101106601
Supplier:  Diversey
New Product
Specifications
  • Description:
    SURE® antibacterial hand wash free, 4×1,3 L, 185×135×283 mm
Specifications
About this item
Soft Care Des E is an alcohol based (71,5%) hand disinfectant gel for waterless skin disinfection. Soft Care Des E is suitable for use in all areas, including healthcare and food processing environments.
Hand disinfectant based on ethanol. Ethanol is a disinfectant which is very effective against resident and transient flora such as bacteria (e.g. MRSA) and yeast. It is also effective against enveloped viruses such as influenza and certain non-enveloped viruses such as Norovirus and Adenovirus.
Contains a skin softener helping the skin to remain smooth and intact, particularly in areas where frequent hand disinfection is required.
Perfume-free eliminating the risk of tainting foods and making it suitable for application in food processing areas.
Skin compatibility dermatologically tested (Patch and TEWL tests.