Order Entry
Slovenia
ContactUsLinkComponent
L-Amino Acid Assay Kit
L-Amino Acid Assay Kit
Catalog # SIALMAK490-1KT
Supplier:  Merck
New Product
CAS Number:  
L-Amino Acid Assay Kit
Catalog # SIALMAK490-1KT
Supplier:  Merck
New Product
CAS Number:  

Specifications

  • Description:
    L-Amino acid assay kit

Specifications

About this item

L-amino acids are essential molecules defined by the nature of their side-chain. There are 20 amino acids that are commonly used by most organisms for the production of proteins and other biologically relevant molecules. While amino acids can occur in either the D- or L-enantimer, only the L-form is used by cells.

  • Linear detection range: 3,3 to 500 µM (colorimetric assay) and 0,13 to 50 µM (fluorometric assay) for 60 minute reaction

    Compatible with high-throughput handling systems

The kit is suitable for L-Amino Acid determination in serum, culture media, tissue homogenates, cell lysates, urine, food samples, etc.

The L-Amino Acid Assay Kit uses an enzyme-catalysed oxidation of L-amino acids to convert a dye into a colored or fluorescent form. The absorbance at 570 nm or fluorescence intensity at λEx = 530 nm/λEm = 585 is directly proportional to the L-amino acid concentration in the sample.

Application(s): Food and beverages
Detection method: Colorimetric, fluorometric
Relevant disease(s): Genitourinary disorders; cancer
Shipped in: Wet ice
Storage temperature: −20 °C