ContactUsLinkComponent
Tork Xpress® H2 Multifold Hand Towels, Blue
Catalog # SCAA150388
Supplier:  Essity
New Product
undefined
Tork Xpress® H2 Multifold Hand Towels, Blue
Catalog # SCAA150388
Supplier:  Essity
New Product
Specifications
  • For:
    Compatible with Tork H2 dispensers
  • Packed:
    20 packs, 237 paper hand towels each; 2 ply
  • Colour:
    Blue
  • Description:
    Universal multifold hand towels, refill
  • L×W:
    78×213 mm (Folded size); 234×213 mm (Unfolded size)
Specifications
About this item
Tork Xpress® multifold hand towels blue H2, universal, 2-ply, Z-fold, economical. Perfectly balance budget and performance with these universal quality multifold hand towels.
  • Reduce consumption and boost hygiene with one-at-a-time dispensing when used with a compatible Tork towel dispenser
  • Commit to a lower environmental impact with these EU Ecolabel certified hand towels
  • Suit the needs of medium-traffic washrooms with these economical paper hand towels
    Universal quality, compatible with H2 dispensers
These practical paper towels meet basic hand-drying requirements while being easy on the budget. The multifold paper towels combine with the Tork Xpress® towel dispenser to offer one-at-a-time dispensing, which encourages guests to only take what they need - reducing hand towel waste and ensuring users get the most out of their money. This also improves the hygiene in medium-traffic washrooms by reducing the chance of cross-contamination.
Certifications:
FSC Recycled credit, SA-COC-008266; EU Ecolabel, SE/004/001.