ContactUsLinkComponent
Tork Xpress® H2 Flushable Multifold Hand Towels, White
Catalog # SCAA129089
Supplier:  Essity
New Product
undefined
Tork Xpress® H2 Flushable Multifold Hand Towels, White
Catalog # SCAA129089
Supplier:  Essity
New Product
Specifications
  • For:
    Compatible with Tork H2 dispensers
  • Packed:
    21 packs, 200 paper hand towels each; 2 ply
  • Colour:
    White
  • Description:
    Advanced multifold hand towels, refill
  • L×W:
    80×213 mm (Folded size); 240×213 mm (Unfolded size)
Specifications
About this item
Tork Xpress® flushable multifold hand towels white H2, advanced, 2-ply, Z-fold, fast dissolving.
  • Make a great impression, thanks to the attractive Tork leaf décor on our multifold paper towels
  • Reduce paper towel consumption in washrooms and improve hygiene with one-at-a-time dispensing
  • Help prevent blockages with these fast-dissolving paper hand towels
  • Maximise the space - use these hand towels with the compact Tork Xpress® dispenser in low to medium-traffic washrooms
  • Advanced quality, compatible with H2 dispensers
Maximise hygiene and really impress guests with the Tork Xpress® flushable multifold hand towels. The fast-dissolving paper hand towels won't cause blockages and, when used in a Tork Xpress® hand towel dispenser, encourage responsible consumption with one-at-a-time dispensing. This also means visitors only touch the towel they use, improving hygiene. Plus, guests will appreciate the paper towel's attractive look and grey leaf embossing.
Certifications:
FSC MIX Credit, SA-COC-008266; EU Ecolabel, SE/004/001.