Thermoshakers for microtubes and PCR plates, PHMT series
444-0340
- Orbit:2 mm
- Speed range:250 - 1400 min⁻¹
- Temperature range:Ambient +5…100 °C
- Description:Thermoshaker, PHMT
- Model:PHMT
- Temperature uniformity:±0,1 °C
- Speed setting:10 min⁻¹
- Heat-up time:10 min
- Weight:4 kg
- W×D×H:205x230x130 mm
- Cat. no.:444-0340
- Pk:1