ContactUsLinkComponent
Shaking and Heating Module Teleshake 95 for Liquid Handling Station
Shaking and Heating Module Teleshake 95 for Liquid Handling Station
Catalog # BRND709470
Supplier:  Brand
undefined
Shaking and Heating Module Teleshake 95 for Liquid Handling Station
Catalog # BRND709470
Supplier:  Brand
Specifications
  • Description:
    Shaking and heating module Teleshake 95 for liquid handling station items supplied heating/shaking module with centering plate, incl. universal flat-bottom adapter plate.
  • Environmentally Preferable:
Specifications
About this item
With the Teleshake 95 module, shaking and heating of samples in microplates and PCR consumables is possible directly inside the Liquid Handling Station pipetting robot from BRAND, without removing the plates from the robot.
  • For shaking and heating 96-well and 384-well microplates and PCR plates
  • Shaking frequency range 100 - 1900 rpm
  • Max temperature 90 °C
Saves time and reduces the risk of sample contamination. Applications include sample incubation and intensive stirring of samples during reaction at room temperature or higher. Thanks to its modular design, installation of the heater/shaker is easy. Simply place it on the worktable of the Liquid Handling Station and connect it via a USB port to a PC or notebook and the included controller.