Order Entry
Slovenia
ContactUsLinkComponent
Human Procollagen III N-terminal propeptide (PIIINP) Antibody Pair - BSA and Azide Free
Human Procollagen III N-terminal propeptide (PIIINP) Antibody Pair - BSA and Azide Free
Catalog # ABCAAB253842-10X96
Supplier:  Abcam
CAS Number:  
undefined
Human Procollagen III N-terminal propeptide (PIIINP) Antibody Pair - BSA and Azide Free
Catalog # ABCAAB253842-10X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Format:
    Uncoated
  • Target protein:
    Procollagen III N-terminal propeptide (PIIINP)
  • Description:
    Human Procollagen III N-terminal propeptide (PIIINP) antibody pair - BSA and azide free
  • Detection range:
    39,1 - 5000 pg/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Tests per kit:
    10×96 tests

Specifications

About this item

Human Procollagen III N-terminal propeptide (PIIINP) antibody pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Procollagen III N-terminal propeptide (PIIINP).