ContactUsLinkComponent
Complex IV Human Enzyme Activity Dipstick Assay Kit
Complex IV Human Enzyme Activity Dipstick Assay Kit
Catalog # ABCAAB109876-30
Supplier:  Abcam
CAS Number:  
undefined
Complex IV Human Enzyme Activity Dipstick Assay Kit
Catalog # ABCAAB109876-30
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Complex IV human enzyme activity dipstick assay kit
  • Method:
    Sandwich
  • Protein:
    Complex IV
  • Species reactivity:
    Other;Human
  • Processing time:
    Multiple steps
  • Product type:
    Pre-coated
  • Tests per kit:
    30 tests
Specifications
About this item
Complex IV Human Enzyme Activity Dipstick Assay Kit is an immunoassay kit for the detection of Complex IV Human Enzyme Activity in Cow, Human in Cell culture extracts, Tissue samples.