uniflex 7020 Assembly Gloves, 60515 Series
Supplier: UVEX
Certificates
About this item
The uniflex 7020 in cotton interlock is very versatile and particularly suitable for oily and wet working environments.
- Protective glove for general activities
- Very versatile
- Good flexibility
- Very good abrasion resistance
The protective glove’s backing material ensures a comfortable fit and flexible movements. The nitrile rubber coating on the back of the glove also ensures good abrasion resistance under mechanical stress, for example during maintenance or machine work.
Specifications
- Catalog No:
- UVEX6051587
- UVEX6051590
- UVEX6051589
- UVEX6051588
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- EN norm:
- EN 420:2003 + A1:2009/EN 388:2016
- EN 420:2003 + A1:2009/EN 388:2016
- EN 420:2003 + A1:2009/EN 388:2016
- EN 420:2003 + A1:2009/EN 388:2016