Anti-Histone H3 (di methyl K4 + tri methyl K4) Mouse Monoclonal Antibody [clone: mAbcam 6000] (Alexa Fluor® 488)
Catalog # ABCAAB205814-100
Supplier: Abcam
Specifications
- Seq:1286
- Size:100 µl
- Pack type:Vial
Specifications
About this item
Alexa Fluor® 488 Mouse monoclonal [mAbcam 6000] to Histone H3 (di methyl K4, tri methyl K4).
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: mAbcam 6000
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human