HPLC columns, Pursuit™ XRsUltra 2,8
: VARIAN
Pursuit™XRs Ultra UPS 2,8 columns are ultra-purity silica based columns.
- Optimised particles for higher efficiency
- Decrease runtime while maintaining resolution
- Reduced backpressure
- Catalog No:
- VARIA7501100X030
- VARIA7501150X020
- VARIA7501100X020
- VARIA7511050X020
- VARIA7511100X020
- VARIA7501150X030
- VARIA7501030X020
- VARIA7511030X020
- VARIA7521050X020
- VARIA7521100X020
- VARIA7511150X030
- VARIA7521030X020
- VARIA7501050X020
- Description:
- C18
- C18
- C18
- C8
- C8
- C18
- C18
- C8
- Diphenyl
- Diphenyl
- C8
- Diphenyl
- C18
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 1,5 - 10
- 1,5 - 10
- 1,5 - 10
- 1,5 - 10
- 1,5 - 10
- 1,5 - 10
- 1,5 - 10
- 1,5 - 10
- 1,5 - 8,0
- 1,5 - 8,0
- 1,5 - 10
- 1,5 - 8,0
- 1,5 - 10
Frequently Bought Together