Molecular model: DNA, Molymod®
Molecular model of DNA with four bases
Catalog # SPIRMKO-ACGT-53
Supplier: SPIRING ENTERPRISES
Jump to:
Product Details & Documents
This molecular model can show how the bases (adenin, cytosin, guanin, thymin) are combined through triple and double hydrogen bonds.
Specifications
- Description:Molecular model of DNA with four bases