ContactUsLinkComponent
Gel loading pipette tips
Gel loading pipette tips
Catalog # GREI770280_960
Supplier:  Greiner Bio-One
Gel loading pipette tips
Catalog # GREI770280_960
Supplier:  Greiner Bio-One
Specifications
  • Packed:
    10 racks, 96 each
  • Sterile:
    -
  • Volume:
    10 - 200 µl
  • Tip style:
    Elongated end
  • Tip colour:
    Transparent
  • For use with:
    Universal
  • Pk:
    960
Specifications
About this item
These gel loading tips are characterised by an elongated end. They are ideal for numerous molecular biological applications, especially for filling the slots of sequencing, PCR and protein electrophoresis gels.
  • Free of detectable DNase, RNase and human DNA
  • Non pyrogenic
  • Non cytotoxic