ContactUsLinkComponent
Urine collection bottles, for women
  391-0024
undefined
Urine collection bottles, for women
  391-0024
  • Capacity:
    1000 ml
  • Pk:
    1
Urinals made of PE for women.
  • Natural coloured
  • Approximate scale, with cover
  • Heat resistant up to 90 °C