Separating funnels, pear shaped
Catalog # HECH42970066
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Socket size:29/32 NS
- Capacity:1000 ml
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Separating funnels, made of borosilicate glass, pear shaped, stopcock with solid glass plug.
- Without graduations
- Excellent chemical resistance
- Excellent temperature resistance