Orders LinkContactUsLinkComponent
Measuring scoops
Measuring scoops
Supplier:   VITLAB
Measuring scoops
Supplier:  VITLAB
Certificates
Measuring scoops
Measuring scoops
8 Options Available Below
Product Details & Documents
PP, coloured. With precision formed filling edge and comfortable, stable handle.
  • Also suitable as weighing scoops
  • Easily readable volume quantities visible on the upper side of the handle
  • Food-safe product according to EC Directive No. 10/2011
Product Family Options
SET
Product Information
  • Capacity
    Colour
    Availability
    Price
  • 231-0562
    Set of 8 coloured scoops (white, red, grey, black, yellow, dark blue, light blue, green)
    Set of 8 coloured scoops (white, red, grey, black, yellow, dark blue, light blue, green)
    100 ml
    Assorted
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 15,40 €
    15,40 €
    1 * 1 items
       15,40 €
Product Information
  • Capacity
    Colour
    Availability
    Price
  • 100 ml
    Yellow
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - 20,60 €
    20,60 €
    1 * 12 items
       20,60 €
  • 100 ml
    Red
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - 20,60 €
    20,60 €
    1 * 12 items
       20,60 €
  • 100 ml
    Grey
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - 20,60 €
    20,60 €
    1 * 12 items
       20,60 €
  • 100 ml
    Black
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - 20,60 €
    20,60 €
    1 * 12 items
       20,60 €
  • 100 ml
    Dark blue
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - 20,60 €
    20,60 €
    1 * 12 items
       20,60 €
  • 100 ml
    Green
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - 20,60 €
    20,60 €
    1 * 12 items
       20,60 €
  • 100 ml
    Light blue
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - 20,60 €
    20,60 €
    1 * 12 items
       20,60 €