ContactUsLinkComponent
Centrifuge tubes, with screw cap
Centrifuge tubes, with screw cap
Catalog # 212-0733
undefined
Centrifuge tubes, with screw cap
Catalog # 212-0733
Specifications
  • Closure type:
    Screw cap
  • Ø×L:
    17×112 mm
  • Thread:
    GL 14
  • Capacity:
    15 ml
  • Pk:
    100
Specifications
About this item
AR®-Glass
  • Ungraduated
  • Good chemical resistance
Conical bottom, with thread GL 14 and screw cap of polypropylene.