Side bin for medium waste baskets
Fits onto medium sized wastebasket for a convenient deskside recycling station, Black, Resins, 5 L, 18,54×26,92×29,72 mm
Catalog # 129-1818
Supplier: Rubbermaid Commercial Products
Jump to:
Product Details & Documents
The Rubbermaid Commercial Recycling Side Bin is constructed of polyethylene to be lightweight and durable. It attaches onto medium wastebaskets to create a deskside recycling solution.
Space-efficient, economical, and an easy and an effective way to recycle
- All-plastic construction won’t chip, rust or dent Rolled rims add strength, and are easy to clean
- Fits onto medium sized wastebasket for a convenient deskside recycling station
Specifications
- Capacity:5 L
- Colour:Black
- Description:Fits onto medium sized wastebasket for a convenient deskside recycling station
- Material:Resins
- W×D×H:18,54×26,92×29,72 mm