ContactUsLinkComponent
Blot module, miniVE™
Blot module, miniVE™
Catalog # 732-1774
Blot module, miniVE™
Catalog # 732-1774
Specifications
  • Capacity:
    4 gels/transfer
  • Description:
    Blot module, miniVE™
  • Seq:
    03
  • Model:
    Blot module, miniVE™
  • Main unit components:
    Includes blot module, 4 dacron sponges, 6 mm, blotter paper–25 sheets
  • Pk:
    1