Strips of 8 PCR tubes with detached cap strips
Catalog # BITX3426.8.X
Supplier: BIOTIX
Specifications
- Packed:5 bags of 25/pack
- Capacity:0,2 ml
- Colour:Clear
- Description:strips of 8 PCR tubes, with detached cap strips
- Pk:125
Specifications
About this item
Made of virgin polypropylene copolymer, clear.
- Sterile or non sterile
- Autoclavable
- Excellent chemical resistance
Specifically designed for PCR, these tubes feature uniform microthin walls for efficient heat transfer and fast cycling times. Tubes and caps are compatible with most leading thermal cyclers.