Single channel pipettes, mechanical, variable volume, PIPETMAN® Neo™
Catalog # 613-2125
Supplier: GILSON
Specifications
- Description:PIPETMAN® Neo™ starter kit incl. P20N / P200N /P1000N pipettes, D200 and D1000 DIAMOND tips, 3 Jimmy™ microtube openers, 3 SINGLE™ pipette holders, 1 Gilson guide to pipetting
- Pk:1 SET
Specifications
About this item
Air-displacement pipette exhibiting a distinct enhancement in comfort and a wide useful volume range with the same robustness, accuracy and precision as PIPETMAN® Classic™.
- Specially engineered springs provide a significant reduction of pipetting and purge forces - up to 50% reduction of pipetting and purge forces on certain models
- Improved clip ejector results in easier daily operation and even more robust system over time
- Long product life and high resistance to aggressive chemicals thanks to stainless steel and PVDF components
- Air-tight seal with unique 'dry seal' technology (grease-free)
- Colour-coded push buttons facilitate identification of the different pipettes
- Tip holder and tip ejector are fully autoclavable to avoid cross-contamination
- PIPETMAN Neo P2N and P10N are equipped with dual position tip-ejector allowing the use of DIAMOND D10 (short tip) and DL10 (long tip)
PIPETMAN Neo P2N and P10N models are ideal for molecular biology techniques (PCR, DNA sequencing, etc.).
PIPETMAN Neo P20N, P100N, P200N, and P1000N models are ideal for small-volume dispensing of aqueous fluids of moderate viscosity and density (molecular biology, immunology, etc.).