Orders LinkContactUsLinkComponent
N,N-Dimethylformamide
Glass bottle, 2,5 L
Catalog # ROTHCP79.1
Flammable
Health
Irritant
Supplier:   Roth Carl
N,N-Dimethylformamide
Catalog # ROTHCP79.1
Supplier:  Roth Carl
CAS Number:  68-12-2
N,N-Dimethylformamide
N,N-Dimethylformamide
Glass bottle, 2,5 L
CAS Number:  68-12-2
Price
134,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
  • CAS Number:68-12-2
  • Formula:HCON(CH₃)₂
  • MDL Number:MFCD00003284
  • Molecular Weight:73.09 g/mol
  • Merck Index:13,03269
  • Boiling Pt:153 °C (1013 hPa)
  • Melting Pt:–61 °C
  • Density:0.949 g/cm³ (20 °C)
  • Flash Pt:57.5 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-679-5
  • UN:2265
  • ADR:3,III
  • REACH:01-2119475605-32
Specifications
Specifications
  • Pack type:
    Glass bottle
  • Pk:
    2,5 L