Dissection probe
Dissection probe, Straight, 14,5 cm, 1 mm
Catalog # MART16-102-14
Supplier: KLS Martin Group
Jump to:
Product Details & Documents
Dissecting probe completely made of 18/8 stainless steel.
Specifications
- Ø:1 mm
- Form:Straight
- Description:Dissection probe
- Length:14,5 cm