ContactUsLinkComponent
Flasks, iodine, with collar, with hollow glass stopper
Flasks, iodine, with collar, with hollow glass stopper
Catalog # WITG2.438.500
Supplier:  witeg Labortechnik
undefined
Flasks, iodine, with collar, with hollow glass stopper
Catalog # WITG2.438.500
Supplier:  witeg Labortechnik
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    500 ml
  • Pk:
    1
Specifications
About this item
DURAN®, borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Volume graduations and labelling area. Hollow glass stopper with extra long stem.