ContactUsLinkComponent
Infusion filter, Sterifix® Neonat
Infusion filter, Sterifix® Neonat
Catalog # BRAU4099257
Supplier:  B. Braun
undefined
Infusion filter, Sterifix® Neonat
Catalog # BRAU4099257
Supplier:  B. Braun
Specifications
  • Membrane:
    0,2 µm
  • Colour:
    Transparent
  • Description:
    Infusion filter with Luer-lock fitting
  • Pk:
    1
Specifications
About this item
Infusion filter with bacteria and particle retention.
  • Reliable air venting regardless of position
  • High throughput and Supor® membrane
  • Pressure resistant to one bar