Pierce™ Horseradish Peroxidase
Catalog # PIER31491
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- CAS No.:9003-99-0
- Species:Horseradish
- Storage conditions:Freezer
- MDL No.:MFCD00071339
- Enzyme name:Peroxidase