Order Entry
Slovenia
ContactUsLinkComponent
 
Organic reference standard, Benz[a]anthracene 1000 µg/ml in methanol
  PROOU-EPA-1072
 :  
Organic reference standard, Benz[a]anthracene 1000 µg/ml in methanol
  PROOU-EPA-1072
 :  

 

  • Pk:
    1
  • CAS No.:
    56-55-3
  • Description:
    Benz[a]anthracene 1,000 µg/ml in methanol
  • Formula:
    C₁₈H₁₂
  • Description:
    Benz[a]anthracene