Orders LinkContactUsLinkComponent
Standard pH electrodes
 :   CONSORT
Standard pH electrodes
 :  CONSORT
Standard pH electrodes
Standard pH electrodes
  • Catalog No:
  • CONSSP10T
  • CONSSP21B
  • CONSSP21T
  • Model:
  • SP10T (+ ATC/temperature (Pt1000)), single junction, sealed
  • SP21B, single junction, refillable
  • SP21T (+ ATC/temperature (Pt1000)), single junction, refillable
Product Family Options
  • Description
    Type
    pH
    Temp.
    Material
    Ø×L
    Connection
    For
  • SP10T (+ ATC/temperature (Pt1000)), single junction, sealed
    SP10T (+ ATC/temperature (Pt1000)), single junction, sealed
    0 - 14
    0...+80 °C
    Epoxy
    12×110 mm
    BNC + 2 banana plugs, 1 m cable
    Field use, production, education, agriculture
    1 * 1 items
    1 * 1 items - 144,00 €
    144,00 €
    1 * 1 items
       144,00 €
    Specifications
    Cat. No.
    CONSSP10T
    Model
    SP10T (+ ATC/temperature (Pt1000)), single junction, sealed
  • SP21B, single junction, refillable
    SP21B, single junction, refillable
    0 - 14
    0...+100 °C
    Glass
    12×110 mm
    BNC, 1 m cable
    Laboratory use, research, production, education, agriculture
    1 * 1 items
    1 * 1 items - 232,00 €
    232,00 €
    1 * 1 items
       232,00 €
    Specifications
    Cat. No.
    CONSSP21B
    Model
    SP21B, single junction, refillable
  • SP21T (+ ATC/temperature (Pt1000)), single junction, refillable
    SP21T (+ ATC/temperature (Pt1000)), single junction, refillable
    0 - 14
    0...+100 °C
    Glass
    12×110 mm
    BNC + 2 banana plugs, 1 m cable
    Laboratory use, research, production, education, agriculture
    1 * 1 items
    1 * 1 items - 259,00 €
    259,00 €
    1 * 1 items
       259,00 €
    Specifications
    Cat. No.
    CONSSP21T
    Model
    SP21T (+ ATC/temperature (Pt1000)), single junction, refillable