Orders LinkContactUsLinkComponent
Accessories for BioSpectrometer®
Accessories for BioSpectrometer®
Eppendorf BioSpectrometer® Reference filter kit, for checking photometric accuracy and wavelength systematic error (according to NIST®).
Catalog # 634-0796
Supplier:   EPPENDORF
Accessories for BioSpectrometer®
Catalog # 634-0796
Supplier:  EPPENDORF
Accessories for BioSpectrometer®
Accessories for BioSpectrometer®
Eppendorf BioSpectrometer® Reference filter kit, for checking photometric accuracy and wavelength systematic error (according to NIST®).
Price
1 963,00 €
Log in for availability
1 * 1 SET
Product Details & Documents
Specifications
  • Description:
    Eppendorf BioSpectrometer® Reference filter kit, for checking photometric accuracy and wavelength systematic error (according to NIST®).