Orders LinkContactUsLinkComponent
Spatulas
Supplier:   COWIE
Spatulas
Supplier:  COWIE
Certificates
Spatulas
5 Options Available Below
Product Details & Documents
Double-ended spatula/scraper combination made of PTFE, with fine chamfered ends.
Product Family Options
Product Information
  • Length
    Blade width
    Colour
    Availability
    Price
  • 120 mm
    12 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 14,50 €
    14,50 €
    1 * 1 items
       14,50 €
  • 150 mm
    18 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 17,70 €
    17,70 €
    1 * 1 items
       17,70 €
  • 180 mm
    20 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 20,90 €
    20,90 €
    1 * 1 items
       20,90 €
  • 210 mm
    25 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 23,70 €
    23,70 €
    1 * 1 items
       23,70 €
  • 240 mm
    30 mm
    White
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 26,30 €
    26,30 €
    1 * 1 items
       26,30 €