Orders LinkContactUsLinkComponent
Operating scissors
Operating scissors
Supplier:   KLS Martin Group
Operating scissors
Supplier:  KLS Martin Group
Certificates
Operating scissors
5 Options Available Below
Product Details & Documents
Operating scissors made of stainless steel. Available straight or curved.
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • Blunt/blunt, straight
    165 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • Sharp/blunt, straight
    165 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • Sharp/blunt, straight
    185 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • Blunt/blunt, curved
    165 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • Sharp/blunt, curved
    165 mm