Stopcock, three-way, acc. to Czako
Catalog # WITG0.268.001
Supplier: witeg Labortechnik
Specifications
- Hole Ø:1,6 mm
- Pk:1
- Spigot:14,5 NS
Specifications
About this item
Stopcocks made of borosilicate glass.
- With capillary side arms
- Solid glass key
- Good chemical and temperature resistance
Certifications: DIN 12563