Cold storage overall, Matterhorn 6402
Catalog # 113-7157
Supplier: SIOEN
Specifications
- Clothing Size:XL
- Closure type:Zip closure
- Garment application:Outdoor
- Gender:Unisex
- Risk area:Cold
- Colour:Blue/dark blue
- Seq:04
- Model:6402 Matterhorn
- Fabric:Polyester, cotton and Isosoft lining
- Sleeve or leg length:Long
- Weight:225 g/m²
- Pk:1
Specifications
About this item
This overall, made of polyester, cotton and Isosoft lining, offers protection from temperatures down to −40 °C. Cold bridges are avoided.
- Fixed quilted lining, knitted wind cuffs in sleeves
- One inside pocket, one patched chest pocket, two patched pockets, pen pocket
- Fleece inside collar, zip closure under flap with press studs
- Ankles adjustable by touch and close fastening, zip closure under flap at leg
- Elasticated waist
With additional protection and lining at shoulders, elbows and knees, stitched seam and high grade extra protective thermal lining.
Weight: 225 g/m²
Certifications: EN 342 (3-2)