Orders LinkContactUsLinkComponent
Work socks, EMSEN
Work socks, EMSEN
Supplier:   FELDTMANN HELMUT
Work socks, EMSEN
Supplier:  FELDTMANN HELMUT
Certificates
Work socks, EMSEN
2 Options Available Below
Product Details & Documents
Super-comfortable socks with thick padding at the most important weight-bearing zones in the toe, ball and heel area.
  • 80% natural fibers
  • Optimal foot climate due to high cotton content
Fabric: 80% cotton/ 12% polyamide/ 8% elasthane
Colour: Black
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-08294
  • 113-08295
  • Model:
  • BW90
  • BW90
  • Fabric:
  • 80% cotton, 12% polyamide and 8% elasthane
  • 80% cotton, 12% polyamide and 8% elasthane
Specifications
Product Family Options
Product Information
  • Colour
    Garment fit
    Shoe size
    Availability
    Price
  • Black
    Regular
    39 - 42
    Log in for availability
    Log in for availability
    1 * 1 PAIR
    1 * 1 PAIR
    Specifications:
    More
    Specifications
    Cat. No.
    113-08294
    Model
    BW90
    Fabric
    80% cotton, 12% polyamide and 8% elasthane
  • Black
    Regular
    43 - 46
    Log in for availability
    Log in for availability
    1 * 1 PAIR
    1 * 1 PAIR
    Specifications:
    More
    Specifications
    Cat. No.
    113-08295
    Model
    BW90
    Fabric
    80% cotton, 12% polyamide and 8% elasthane