Supplier: RENTEX
Certificates
About this item
Specifications
- Catalog No:
- 113-07044
- 113-05891
- 113-07045
- 113-05886
- 113-05884
- 113-05883
- 113-07037
- 113-05888
- 113-07036
- 113-05887
- 113-07039
- 113-05890
- 113-07038
- 113-05889
- 113-07040
- 113-05892
- 113-07042
- 113-07041
- 113-05821
- 113-05820
- 113-07043
- 113-05813
- 113-05824
- 113-05812
- 113-05815
- 113-05822
- 113-05814
- 113-05823
- 113-05817
- 113-05816
- 113-05819
- 113-05818
- 113-07035
- 113-05885
- Gender:
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- For ladies
- Sleeve or leg length:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- 65% Polyester, 35% Baumwolle
- Weight:
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²
- 215 g/m²