Overalls, chemical protection, decontex® Concept
Supplier: KIND ARBEITSSICHERHEIT
Certificates
About this item
Flame retardant and anti-static overalls made from elastomeric special PVC.
- Integrated elasticated hood, front zip fastening with rip-tape cover for optimum fit
- Additional internal chest panel for increased safety, especially for protection against more intense exposure to liquid media
- Protective bib at the top, elasticated band
- Sleeve ends with internal safety closures, split at the ankle with rip-tape straps making them more comfortable to pull on over clothes
Ideal for general chemical protection, cleaning, decontamination and disposal, transport (hazardous goods), tank cleaning.
Certifications: EN 14605 (Type 4), EN 1149-1, EN ISO 14116 Index 1
Specifications
- Catalog No:
- 113-08355
- 113-08354
- 113-08353
- 113-08352
- Model:
- Concept
- Concept
- Concept
- Concept
- Anti-static:
- +
- +
- +
- +
- Garment application:
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial