Shoe covers, low profile, chemical resistant, Cleansafe® 2
113-08388
: TRENKA
- Garment application:Chemical protection,Laboratory
- Colour:White
- Pk:1 PAIR
Disposable shoe covers made from PP/PE, to fit all shoe sizes. Elasticated.
Height: Approx. 15 cm, Length: 34 cm
: EN 13034 (Type 6); EN 13982 (Type 5); EN 1149-1