ContactUsLinkComponent
Shoe covers, low profile, chemical resistant, Cleansafe® 2
  113-08388
 :  TRENKA
undefined
Shoe covers, low profile, chemical resistant, Cleansafe® 2
  113-08388
 :  TRENKA
  • Garment application:
    Chemical protection,
    Laboratory
  • Colour:
    White
  • Pk:
    1 PAIR
Disposable shoe covers made from PP/PE, to fit all shoe sizes. Elasticated.
Height: Approx. 15 cm, Length: 34 cm
 : EN 13034 (Type 6); EN 13982 (Type 5); EN 1149-1