Overall, Cleansafe® 1
Catalog # 113-08351
Supplier: TRENKA
Specifications
- Clothing Size:XXL
- Anti-static:+
- Garment application:Chemical protection,General Purpose,Industrial
- Colour:White
- Pk:1
Specifications
About this item
Superb™ disposable coveralls specifically designed for disposal work.
- Front zip with zip flap
- Attached socks and elasticated hood, hips and wrists
- Excellent design that finishes above the chin avoiding exposed neck areas
Größe: XXL
Schutzkleidung Typ 5+6
Certifications: Typ 5+6